missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Alanine-3,3,3-d3-N-t-BOC, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | N-(tert-Butoxycarbonyl)-L-alanine-3,3,3-D3 |
| CAS | 161602-47-7 |
| Molecular Formula | C8 D3 H12 N O4 |
| Quantity | 0.5 g |
| Synonym | L-Alanine-3,3,3-d3, N-[(1,1-dimethylethoxy)carbonyl]- (9 CI, ACI), N-[(1,1-Dimethylethoxy)carbonyl]-L-alanine-3,3,3-d3 (ACI), (S)-2-((tert-Butoxycarbonyl)amino)propanoic-3,3,3-d3 acid, (2 S)-2-(tert-Butoxycarbonylamino)propionic-3,3,3-d3 acid, (2 S)-2-[(2-Methylpropan-2-yl)oxycarbonylamino]propanoic-3,3,3-d3 acid, N-BOC-L-alanine-3,3,3-d3, N-tert-Butoxycarbonyl-L-alanine-3,3,3-d3, N-(tert-Butoxycarbonyl)-L-alanine-3,3,3-D3, N-tert-Boc-L-alanine-3,3,3-d3, L-Alanine-3,3,3-d3-N-t-BOC |
| InChI Formula | InChI=1 S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])[C@H](NC(=O)OC(C)(C)C)C(=O)O |
| IUPAC Name | (2 S)-3,3,3-trideuterio-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| Molecular Weight (g/mol) | 192.2275 |
| Formula Weight | 192.1189 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction