Learn More
p-Toluidine-d9, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 4-Methylaniline D9 |
| CAS | 352431-23-3 |
| Molecular Formula | C7 D9 N |
| Quantity | 0.5 g |
| Synonym | Benzen-2,3,5,6-d4-amine-d2, 4-(methyl-d3)- (9 CI), 4-(Methyl-d3)benzen-2,3,5,6-d4-amine-d2, 4-Toluidine-d9, p-Toluidine-d9, 4-Methylaniline-D9, 4-Methylaniline D9, p-Toluidine D9 |
| InChI Formula | InChI=1 S/C7H9N/c1-6-2-4-7(8)5-3-6/h2-5 H,8H2,1H3/i1D3,2 D,3 D,4 D,5 D/hD2 |
| SMILES | [2 H]N([2 H])c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N,N,2,3,5,6-hexadeuterio-4-(trideuteriomethyl)aniline |
| Molecular Weight (g/mol) | 116.209 |
| Formula Weight | 116.13 g/mol |
| Show More |