missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperazine-3,3,5,5-d4-N-t-BOC, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | N-Boc-piperazine-3,3,5,5-D4 |
| CAS | 1126621-87-1 |
| Molecular Formula | C9 D4 H14 N2 O2 |
| Quantity | 0.5 g |
| Synonym | 1-Piperazine-3,3,5,5-d4-carboxylic acid, 1,1-dimethylethyl ester (ACI), 1,1-Dimethylethyl 1-piperazine-3,3,5,5-d4-carboxylate (ACI), N-Boc-piperazine-d4, Piperazine-3,3,5,5-d4-N-t-BOC |
| InChI Formula | InChI=1 S/C9H18N2O2/c1-9(2,3)13-8(12)11-6-4-10-5-7-11/h10H,4-7H2,1-3H3/i4D2,5D2 |
| SMILES | [2 H]C1([2 H])CN(CC([2 H])([2 H])N1)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl 3,3,5,5-tetradeuteriopiperazine-1-carboxylate |
| Molecular Weight (g/mol) | 190.2761 |
| Formula Weight | 190.1619 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction