missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic Anhydride-d10, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Propionic Anhydride-d10 |
| CAS | 870471-31-1 |
| Molecular Formula | C6 D10 O3 |
| Quantity | 0.5 g |
| Synonym | 1,1'-Anhydride Propanoic Acid-d10, Anhydride Propanoic Acid-d10, Methylacetic Anhydride-d10, Propanoic Anhydride-d10, Propionic Acid Anhydride-d10, Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI), Propanoic-d5 acid, anhydride (9 CI) |
| InChI Formula | InChI=1 S/C6H10O3/c1-3-5(7)9-6(8)4-2/h3-4H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C(=O)OC(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,3-pentadeuteriopropanoyl 2,2,3,3,3-pentadeuteriopropanoate |
| Molecular Weight (g/mol) | 140.2 |
| Formula Weight | 140.1258 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction