missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoro-2-propanol-d2, TRC

Product Code. 30392245
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392245 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392245 Supplier Toronto Research Chemicals Supplier No. TRCH293882500MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,1,3,3,3-Hexafluoro-2-propanol-d2
CAS 38701-74-5
Molecular Formula C3D2F6O
Quantity 500 mg
Synonym 1,1,1,3,3,3-Hexafluoro-2-hydroxypropane-d2, 1,1,1,3,3,3-Hexafluoroisopropanol-d2, 1,1,1,3,3,3-Hexafluoroisopropyl Alcohol-d2, 1,1,1,3,3,3-Hexafluoropropanol-d2, 2,2,2-Trifluoro-1-(trifluoromethyl)ethanol-d2, Bis(trifluoromethyl)methanol-d2, HFIP-d2, Hexafluoroisopropanol-d2, Hexafluoroisopropyl Alcohol-d2, NSC 96336-d2, USP Sevoflurane Related Compound C-d2
InChI Formula InChI=1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H/i1D,10D
SMILES [2H]OC([2H])(C(F)(F)F)C(F)(F)F
IUPAC Name 2-deuterio-2-deuteriooxy-1,1,1,3,3,3-hexafluoropropane
Molecular Weight (g/mol) 170.05
Formula Weight 170.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.