missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(1,3-Phenylene)bis-1H-1,2,4-triazole, TRC

Product Code. 30138219
Change view
Click to view available options
Quantity:
5 mg
50 mg
250 mg
Unit Size:
250mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138219 5 mg 5mg
30135560 250 mg 250mg
17944725 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138219 Supplier Toronto Research Chemicals Supplier No. TRCP3198305MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 514222-44-7
Molecular Formula C10 H8 N6
Molecular Weight (g/mol) 212.21
Synonym 1,1'-(1,3-Phenylene)di-1H-1,2,4-triazole, Fluconazole Imp. C (EP)
IUPAC Name 1-[3-(1,2,4-triazol-1-yl)phenyl]-1,2,4-triazole
SMILES c1cc(cc(c1)n2cncn2)n3cncn3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H8N6/c1-2-9(15-7-11-5-13-15)4-10(3-1)16-8-12-6-14-16/h1-8H
Formula Weight 212.081
Percent Purity >95%
Chemical Name or Material 1,1'-(1,3-phenylene)di-1H-1,2,4-triazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.