missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Kestopentaose, TRC

Product Code. 30134622
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134622 100 mg 100mg
30133016 50 mg 50mg
30008314 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134622 Supplier Toronto Research Chemicals Supplier No. TRCK264853100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59432-60-9
Molecular Formula C30 H52 O26
Molecular Weight (g/mol) 828.72
Synonym α-D-Glucopyranoside, O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)- (6CI), O-β-D-Fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)-O-β-D-fructofuranosyl-(2→1)-β-D-fructofuranosyl α-D-glucopyranoside, 1,1,1-Kestopentaose, 1-Kestopentaose, 1F-(1-β-Fructofuranosyl)3-sucrose, 1F-Fructofuranosylnystose, 1F-Fructosylnystose, Fructofuranosyl nystose, Fructosyl nystose, GF4
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-2-[[(2R,3S,4S,5R)-2-[[(2R,3S,4S,5R)-2-[[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxymethyl]-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@]2(CO[C@]3(CO[C@]4(CO[C@]5(CO)O[C@H](CO)[C@@H](O)[C@@H]5O)O[C@H](CO)[C@@H](O)[C@@H]4O)O[C@H](CO)[C@@H](O)[C@@H]3O)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C30H52O26/c31-1-10-15(37)20(42)21(43)26(51-10)56-30(25(47)19(41)14(5-35)55-30)9-50-29(24(46)18(40)13(4-34)54-29)8-49-28(23(45)17(39)12(3-33)53-28)7-48-27(6-36)22(44)16(38)11(2-32)52-27/h10-26,31-47H,1-9H2/t10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20+,21-,22+,23+,24+,25+,26-,27-,28-,29-,30+/m1/s1
Formula Weight 828.2747
Percent Purity >95%
Chemical Name or Material 1,1,1-Kestopentaose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.