missing translation for 'onlineSavingsMsg'
Learn More

1,1',1' '-Methylidynetris[4-isocyanato-benzene (25-35% in ethyl acetate) (>85%), TRC

Product Code. 30010677
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010677 5 g 5g
30004894 2.5 g 2.5g
30007460 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010677 Supplier Toronto Research Chemicals Supplier No. TRCM3124605G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2422-91-5
Molecular Formula C22H13N3O3
Molecular Weight (g/mol) 367.36
IUPAC Name 1-[bis(4-isocyanatophenyl)methyl]-4-isocyanatobenzene
SMILES O=C=Nc1ccc(cc1)C(c2ccc(cc2)N=C=O)c3ccc(cc3)N=C=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C22H13N3O3/c26-13-23-19-7-1-16(2-8-19)22(17-3-9-20(10-4-17)24-14-27)18-5-11-21(12-6-18)25-15-28/h1-12,22H
Formula Weight 367.0957
Concentration or Composition (by Analyte or Components) 1,1',1' '-Methylidynetris-4-isocyanato-benzene 25-35 %
Chemical Name or Material 1,1',1' '-Methylidynetris-4-isocyanato-benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.