missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-1-butanesulfonamide, TRC

Product Code. 30133084
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133084 500 mg 500mg
30011894 50 mg 50mg
30012574 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133084 Supplier Toronto Research Chemicals Supplier No. TRCN649505500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34454-99-4
Molecular Formula C6H6F9NO3S
Molecular Weight (g/mol) 343.17
Synonym N(2-Hydroxyethyl)perfluorobutanesulfonamide, N-(2-Hydroxyethyl)-1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonamide;
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N-(2-hydroxyethyl)butane-1-sulfonamide
SMILES OCCNS(=O)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H6F9NO3S/c7-3(8,5(11,12)13)4(9,10)6(14,15)20(18,19)16-1-2-17/h16-17H,1-2H2
Formula Weight 342.9925
Chemical Name or Material 1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-1-butanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.