missing translation for 'onlineSavingsMsg'
Learn More

1,1':3',1″-Terphenyl-5'-boronic acid, 95%

Product Code. 11486600
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486600 5 g 5g
15483737 250 mg 250mg
11476600 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486600 Supplier Thermo Scientific Alfa Aesar Supplier No. H27640.06
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 128388-54-5 | C18H15BO2 | 274.126 g/mol

A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls. Good chemoselectivity was observed. Unsymmetrically substituted triphenylenes were also easily prepared. A double Suzuki cross-coupling protocol has been devised as a practical route to a variety of terphenyls and triphenylenes.

Solubility
Insoluble in water.

Notes
Refrigerated (2-4°C). Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128388-54-5
Molecular Formula C18H15BO2
Molecular Weight (g/mol) 274.126
MDL Number MFCD09953491
InChI Key MRBZYVMZUBUDAX-UHFFFAOYSA-N
Synonym 3,5-diphenylphenyl boronic acid, 1,1':3',1-terphenyl-5'-ylboronic acid, 1,1':3',1-terphenyl-5'-boronic acid, 3,5-diphenylbenzeneboronic acid, 5'-m-terphenylboronic acid, boronic acid, 1,1':3',1-terphenyl-5'-yl, boronic acid,b-1,1':3',1-terphenyl-5'-yl, 5-phenyl-1,1'-biphenyl-3-ylboronic acid, 3,5-diphenylphenyl, acmc-1cc4x
PubChem CID 14739363
IUPAC Name (3,5-diphenylphenyl)boronic acid
SMILES B(C1=CC(=CC(=C1)C2=CC=CC=C2)C3=CC=CC=C3)(O)O

Specifications

Melting Point 292°C to 296°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 274.13
Percent Purity 95%
Chemical Name or Material 1,1':3',1''-Terphenyl-5'-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.