missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,4,6-Pentamethyl-1,2,3,4-tetrahydronaphthalene, TRC

Product Code. 30423767
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423767 10 g 10g
30417699 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30423767 Supplier Toronto Research Chemicals Supplier No. TRCP27176510G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6683-48-3
Molecular Formula C15 H22
Molecular Weight (g/mol) 202.34
Synonym Bexarotene Impurity 20
IUPAC Name 1,1,4,4,6-pentamethyl-2,3-dihydronaphthalene
SMILES Cc1ccc2c(c1)C(C)(C)CCC2(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H22/c1-11-6-7-12-13(10-11)15(4,5)9-8-14(12,2)3/h6-7,10H,8-9H2,1-5H3
Formula Weight 202.1722
Chemical Name or Material 1,1,4,4,6-Pentamethyl-1,2,3,4-tetrahydronaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.