missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic acid, 99%

Product Code. 11472447
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472447 5 g 5g
11482447 25 g 25g
11424250 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472447 Supplier Thermo Scientific Alfa Aesar Supplier No. A14288.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5445-51-2 | C6H8O4 | 144.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5445-51-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00001325
InChI Key CCQPAEQGAVNNIA-UHFFFAOYSA-N
Synonym 1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
PubChem CID 2568
ChEBI CHEBI:35691
IUPAC Name cyclobutane-1,1-dicarboxylic acid
SMILES C1CC(C1)(C(=O)O)C(=O)O

Specifications

Melting Point ∼158°C (decomposition)
Quantity 5 g
UN Number UN3261
Beilstein 2046031
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material 1,1-Cyclobutanedicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.