missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic Acid, TRC

Product Code. 30136142
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136142 10 g 10g
30134941 100 g 100g
30005964 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136142 Supplier Toronto Research Chemicals Supplier No. TRCB41650310G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5445-51-2
Molecular Formula C6 H8 O4
Molecular Weight (g/mol) 144.13
Synonym 1,1-Cyclobutanedicarboxylic acid, 1,1-Dicarboxycyclobutane, 1-Carboxycyclobutanecarboxylic acid, NSC 22073, Carboplatin Imp. B (EP), Cyclobutane-1,1-dicarboxylic Acid
IUPAC Name cyclobutane-1,1-dicarboxylic acid
SMILES OC(=O)C1(CCC1)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H8O4/c7-4(8)6(5(9)10)2-1-3-6/h1-3H2,(H,7,8)(H,9,10)
Formula Weight 144.0423
Percent Purity >95%
Chemical Name or Material 1,1-Cyclobutanedicarboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.