missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclopropanedicarboxylic acid monomethyl ester, 97%

Product Code. 11415550
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415550 1 g 1g
11425550 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415550 Supplier Thermo Scientific Alfa Aesar Supplier No. H25828.03
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 113020-21-6 | C6H8O4 | 144.126 g/mol

1,1-Cyclopropanedicarboxylic acid monomethyl ester is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Cyclopropanedicarboxylic acid monomethyl ester is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113020-21-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD06800496
InChI Key IZYOHLOUZVEIOS-UHFFFAOYSA-N
Synonym 1-methoxycarbonyl cyclopropanecarboxylic acid, 1,1-cyclopropanedicarboxylic acid monomethyl ester, 1-methoxycarbonyl cyclopropane-1-carboxylic acid, cyclopropane-1,1-dicarboxylic acid, methyl ester, 1-methoxycarbonylcyclopropanecarboxylic acid, 1,1-cyclopropanedicarboxylic acid-1-methyl ester, 1-methoxycarbonyl cyclopropanecarboxylicacid, acmc-1c7y7, methyl cyclopropane-1,1-dicarboxylate, 1-methoxycarbonylcyclopropane carboxylic acid
PubChem CID 13861823
IUPAC Name 1-methoxycarbonylcyclopropane-1-carboxylic acid
SMILES COC(=O)C1(CC1)C(=O)O

Specifications

Melting Point 50°C to 57°C
Boiling Point 85°C to 87°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 144.13
Percent Purity 97%
Chemical Name or Material 1,1-Cyclopropanedicarboxylic acid monomethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.