missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethoxyethane, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1,1-Dimethoxyethane |
| CAS | 534-15-6 |
| Molecular Formula | C4H10O2 |
| Quantity | 25 g |
| Synonym | Acetaldehyde, dimethyl acetal (6CI,8CI), 3-Methyl-2,4-dioxapentane, Acetaldehyde methyl acetal, Dimethyl acetal, Ethylidene dimethyl ether |
| InChI Formula | InChI=1S/C4H10O2/c1-4(5-2)6-3/h4H,1-3H3 |
| SMILES | COC(C)OC |
| Molecular Weight (g/mol) | 90.12 |
| Formula Weight | 90.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction