missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethyloxirane, TRC

Product Code. 30385039
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385039 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385039 Supplier Toronto Research Chemicals Supplier No. TRCD479065100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dimethyloxirane
CAS 558-30-5
Molecular Formula C4H8O
Quantity 100 g
Synonym alpha,alpha-Dimethylethylene Oxide, 2-Epoxy-2-methyl-propane, 1,2-Epoxy-2-methylpropane, 1,2-Epoxyisobutane, 1,2-Isobutylene Oxide, 2,2-Dimethyloxirane, 2-Methyl-1,2-epoxypropane, 2-Methyl-1-propene oxide, 2-Methyl-1-propenoxide, 2-Methylpropylene oxide, Isobutene Oxide, Isobutylene Epoxide, Isobutylene Oxide, NSC 24249
InChI Formula InChI=1S/C4H8O/c1-4(2)3-5-4/h3H2,1-2H3
SMILES CC1(C)CO1
IUPAC Name 2,2-dimethyloxirane
Molecular Weight (g/mol) 72.11
Formula Weight 72.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.