missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dioctadecyl-3,3,3,3-tetramethylindotricarbocyanine Iodide (>90%), TRC

Product Code. 30341994
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30341994 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341994 Supplier Toronto Research Chemicals Supplier No. TRCB41515825MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Dioctadecyl-3,3,3,3-tetramethylindotricarbocyanine Iodide
CAS 100068-60-8
Molecular Formula C63H101IN2
Quantity 25 mg
Synonym DiIC18(7), DiR, XenoLight DiR, 2-[7-(1,3-Dihydro-3,3-dimethyl-1-octadecyl-2H-indol-2-ylidene)-1,3,5-heptatrienyl]-3,3-dimethyl-1-octadecyl-3H-indolium Iodide
InChI Formula InChI=1S/C63H101N2.HI/c1-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38-46-54-64-58-50-44-42-48-56(58)62(3,4)60(64)52-40-36-35-37-41-53-61-63(5,6)57-49-43-45-51-59(57)65(61)55-47-39-34-32-30-28-26-24-22-20-18-16-14-12-10-8-2;/h35-37,40-45,48-53H,7-34,38-39,46-47,54-55H2,1-6H3;1H/q+1;/p-1
SMILES [I-].CCCCCCCCCCCCCCCCCCN1\C(=C/C=C/C=C/C=C/C2=[N+](CCCCCCCCCCCCCCCCCC)c3ccccc3C2(C)C)\C(C)(C)c4ccccc14
IUPAC Name (2Z)-2-[(2E,4E,6E)-7-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)hepta-2,4,6-trienylidene]-3,3-dimethyl-1-octadecylindole;iodide
Molecular Weight (g/mol) 1013.39
Formula Weight 1012.7009
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.