Learn More
1,1'-(Methylenedi-4,1-phenylene)bishydrazine Dihydrochloride, TRC
Product Code. 30468435
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468435 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30468435 Supplier Toronto Research Chemicals Supplier No. TRCM303060100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1'-(Methylenedi-4,1-phenylene)bishydrazine Dihydrochloride
CAS 100829-65-0
Molecular Formula C13H18Cl2N4
Quantity 100 mg
Synonym 1,1'-(Methylenedi-p-phenylene)dihydrazine DiHydrochloride, 4,4'-Dihydrazinodiphenylmethane Hydrochloride
InChI Formula InChI=1S/C13H16N4.2ClH/c14-16-12-5-1-10(2-6-12)9-11-3-7-13(17-15)8-4-11;;/h1-8,16-17H,9,14-15H2;2*1H
SMILES Cl.Cl.NNc1ccc(Cc2ccc(NN)cc2)cc1
IUPAC Name [4-[(4-hydrazinylphenyl)methyl]phenyl]hydrazine;dihydrochloride
Molecular Weight (g/mol) 301.24
Formula Weight 300.0909