missing translation for 'onlineSavingsMsg'
Learn More

1-[10-[2-(Dimethylamino)propyl]phenothiazin-2-yl-1-propanone (>90%), TRC

Product Code. 30135907
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135907 50 mg 50mg
30011899 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135907 Supplier Toronto Research Chemicals Supplier No. TRCD47106550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362-29-8
Molecular Formula C20 H24 N2 O S
Molecular Weight (g/mol) 340.48
Synonym 1-Propanone, 1-[10-[2-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, 1-Propanone, 1-[10-[2-(dimethylamino)propyl]phenothiazin-2-yl]- (6CI,7CI,8CI), 1-[10-[2-(Dimethylamino)propyl]-10H-phenothiazin-2-yl]-1-propanone, (±)-Propiomazine, 1-[10-[2-(Dimethylamino)propyl]phenothiazin-2-yl]-1-propanone, 2-Propionyl-10-[2-(dimethylamino)propyl]phenothiazine, Propiomazine, Propionylpromethazine, Wy 1359
IUPAC Name 1-[10-[2-(dimethylamino)propyl]phenothiazin-2-yl]propan-1-one
SMILES CCC(=O)c1ccc2Sc3ccccc3N(CC(C)N(C)C)c2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H24N2OS/c1-5-18(23)15-10-11-20-17(12-15)22(13-14(2)21(3)4)16-8-6-7-9-19(16)24-20/h6-12,14H,5,13H2,1-4H3
Formula Weight 340.1609
Chemical Name or Material Propionylpromethazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.