missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Maleimide, TRC

Product Code. 30428598
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428598 100 mg -
30428258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428598 Supplier Toronto Research Chemicals Supplier No. TRCP295055100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351870-31-0
Molecular Formula C16H9N3O2
Molecular Weight (g/mol) 275.26
Synonym 1-(1,10-Phenanthrolin-5-yl)-1H-pyrrole-2,5-dione
IUPAC Name 1-(1,10-phenanthrolin-5-yl)pyrrole-2,5-dione
SMILES C1=CC2=CC(=C3C=CC=NC3=C2N=C1)N4C(=O)C=CC4=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H9N3O2/c20-13-5-6-14(21)19(13)12-9-10-3-1-7-17-15(10)16-11(12)4-2-8-18-16/h1-9H
Formula Weight 275.0695
Chemical Name or Material 1,10-Phenanthroline Maleimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.