missing translation for 'onlineSavingsMsg'
Learn More

1,14-Tetradecanediol, TRC

Product Code. 30006934
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006934 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30006934 Supplier Toronto Research Chemicals Supplier No. TRCT291626100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 19812-64-7
Molecular Formula C14H30O2
Quantity 100 mg
Synonym 1,14-Dihydroxytetradecane
InChI Formula InChI=1S/C14H30O2/c15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16/h15-16H,1-14H2
SMILES OCCCCCCCCCCCCCCO
IUPAC Name tetradecane-1,14-diol
Molecular Weight (g/mol) 230.39
Formula Weight 230.2246
Chemical Name or Material 1,14-Tetradecanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.