missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5-Pentabromo-6-(2,3,4,5-tetrabromophenoxy)benzene, TRC

Product Code. 18091090
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18091090 10 mg 10mg
18084000 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18091090 Supplier Toronto Research Chemicals Supplier No. TRCP23786010MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63387-28-0
Molecular Formula C12 H Br9 O
Molecular Weight (g/mol) 880.27
Synonym Benzene, 1,2,3,4,5-pentabromo-6-(2,3,4,5-tetrabromophenoxy)-, Benzene, pentabromo(2,3,4,5-tetrabromophenoxy)- (9CI), 1,2,3,4,5-Pentabromo-6-(2,3,4,5-tetrabromophenoxy)benzene, 2,2',3,3',4,4',5,5',6-Nonabromodiphenyl ether, BDE 206, PBDE 206
IUPAC Name 1,2,3,4,5-pentabromo-6-(2,3,4,5-tetrabromophenoxy)benzene
SMILES Brc1cc(Oc2c(Br)c(Br)c(Br)c(Br)c2Br)c(Br)c(Br)c1Br

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12HBr9O/c13-2-1-3(5(15)6(16)4(2)14)22-12-10(20)8(18)7(17)9(19)11(12)21/h1H
Formula Weight 871.2678
Percent Purity >95%
Chemical Name or Material PBDE 206 (2,2',3,3',4,4',5,5',6-Nonabromodiphenyl Ether)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.