missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4,5-Pentamethylcyclopentadiene, 94%

Product Code. 11323096
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323096 5 g 5g
11313096 1 g 1g
11333096 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323096 Supplier Thermo Scientific Alfa Aesar Supplier No. L10957.06
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4045-44-7 | C10H16 | 136.238 g/mol

1,2,3,4,5-Pentamethylcyclopentadiene serves as a precursor to the ligand 1,2,3,4,5-pentamethylcyclopentadienyl. It is used as a growth modifier chemical, during metal organic chemical vapor deposition of iron from iron pentacarbonyl. It is used as a ligand in one-pot iridium-catalyzed transformation of alcohols to amides. Further, it acts as an electron mediator in the regeneration process of nicotinamide adenine dinucleotide. In addition to this, it undergoes radical cation catalyzed cycloaddition reaction with allenes to get Diels-Alder product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4,5-Pentamethylcyclopentadiene serves as a precursor to the ligand 1,2,3,4,5-pentamethylcyclopentadienyl. It is used as a growth modifier chemical, during metal organic chemical vapor deposition of iron from iron pentacarbonyl. It is used as a ligand in ″one-pot″ iridium-catalyzed transformation of alcohols to amides. Further, it acts as an electron mediator in the regeneration process of nicotinamide adenine dinucleotide. In addition to this, it undergoes radical cation catalyzed cycloaddition reaction with allenes to get Diels-Alder product.

Solubility
Miscible with methanol. dichloromethane and ethyl acetate. Slightly miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4045-44-7
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00001354
InChI Key WQIQNKQYEUMPBM-UHFFFAOYSA-N
Synonym 1,2,3,4,5-pentamethylcyclopentadiene, pentamethylcyclopentadiene, cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, 1,2,3,4,5-pentamethyl, 1,3-cyclopentadiene, pentamethyl, 1,2,3,4,5-pentamethyl-1,3-cyclopentadiene, pubchem20101, acmc-209je4, wqiqnkqyeumpbm-uhfffaoysa, 1,2,3,4,5-pentamethylcyclopentadien
PubChem CID 77667
IUPAC Name 1,2,3,4,5-pentamethylcyclopenta-1,3-diene
SMILES CC1C(=C(C(=C1C)C)C)C

Specifications

Density 0.87
Boiling Point 57°C to 58°C (13 mmHg)
Flash Point 44°C (111°F)
Odor Light
Refractive Index 1.474
Quantity 5 g
UN Number UN3295
Beilstein 1849832
Sensitivity Light sensitive
Solubility Information Miscible with methanol. dichloromethane and ethyl acetate. Slightly miscible with water.
Formula Weight 136.24
Percent Purity 94%
Chemical Name or Material 1,2,3,4,5-Pentamethylcyclopentadiene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.