missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,6-Tetrahydrophthalic Anhydride-3,3,4,5,6,6-D6 (Technical Grade), TRC

Product Code. 30416490
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30416490 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416490 Supplier Toronto Research Chemicals Supplier No. TRCT29417710MG
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,3,6-Tetrahydrophthalic Anhydride-3,3,4,5,6,6-d6
CAS 89614-23-3
Molecular Formula C8 2H6 H2 O3
Quantity 10 mg
Synonym 3a,4,7,7a-Tetrahydro-4,7-d2-1,3-isobenzofurandione-4,5,6,7-d4 (9Cl)
InChI Formula InChI=1S/C8H8O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-2,5-6H,3-4H2/i1D,2D,3D2,4D2
SMILES [2H]C1=C([2H])C([2H])([2H])C2C(C(=O)OC2=O)C1([2H])[2H]
IUPAC Name 4,4,5,6,7,7-hexadeuterio-3a,7a-dihydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol) 158.18
Formula Weight 158.085
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.