Learn More
1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6, TRC
Product Code. 30417927
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417927 10 mg 10mg
30415949 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30417927 Supplier Toronto Research Chemicals Supplier No. TRCT29425010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-d6
CAS 1020719-96-3
Purity Grade Notes HPLC
Molecular Formula C8 2H6 H3 N O2
Quantity 10 mg
Synonym 1H-Isoindole-1,3(2H)-dione-4,5,6,7-d4','3a,4,7,7a-tetrahydro-4,7-d2-, 3a,4,7,7a-Tetrahydro-1H-isoindole-1,3(2H)-dione-4,4,5,6,7,7-d6, 1,2,3,6-Tetrahydrophthalimide-d6, 1,2,3,6-Tetrahydrophthalimide-3,3,4,5,6,6-D6, Δ4-Tetrahydrophthalimide-3,3,4,5,6,6-d6, 3a,4,7,7a-Tetrahydro-4,7-d2-1H-isoindole-1,3(2H)-dione-4,5,6,7-d4 (ACI)
InChI Formula InChI=1S/C8H9NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-2,5-6H,3-4H2,(H,9,10,11)/i1D,2D,3D2,4D2
SMILES [2H]C1=C([2H])C([2H])([2H])C2C(C(=O)NC2=O)C1([2H])[2H]
IUPAC Name 4,4,5,6,7,7-hexadeuterio-3a,7a-dihydroisoindole-1,3-dione
Molecular Weight (g/mol) 157.2
Formula Weight 157.101
Percent Purity >95