missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Tribromopropane, 98%

Product Code. 11336646
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11336646

Brand: Thermo Scientific Alfa Aesar L03752.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 96-11-7 | C3H5Br3 | 280.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-11-7
Molecular Formula C3H5Br3
Molecular Weight (g/mol) 280.79
MDL Number MFCD00017884
InChI Key FHCLGDLYRUPKAM-UHFFFAOYSA-N
Synonym s-tribromopropane, glyceryl tribromohydrin, sym-tribromopropane, glycerol tribromohydrin, propane, 1,2,3-tribromo, unii-d2r8l96tov, ccris 6706, d2r8l96tov, 1,3-tribromopropane, propane,2,3-tribromo
PubChem CID 7279
ChEBI CHEBI:18859
IUPAC Name 1,2,3-tribromopropane
SMILES BrCC(Br)CBr

Specifications

Melting Point 16°C to 17°C
Density 2.398
Boiling Point 220°C
Flash Point 93°C (199°F)
Refractive Index 1.585
Quantity 100 g
Beilstein 1732082
Merck Index 14,9614
Formula Weight 280.8
Percent Purity 98%
Chemical Name or Material 1,2,3-Tribromopropane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.