missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluoro-5-nitrobenzene, 99%

Product Code. 11309166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11309166 5 g 5g
11319166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309166 Supplier Thermo Scientific Alfa Aesar Supplier No. L20302.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 66684-58-0 | C6H2F3NO2 | 177.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66684-58-0
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00456803
InChI Key PTTUMBGORBMNBN-UHFFFAOYSA-N
Synonym 3,4,5-trifluoronitrobenzene, 1-nitro-3,4,5-trifluorobenzene, 3,4,5-trifluoro-nitrobenzene, 1,2,3-trifluoro-5-nitro-benzene, benzene, 1,2,3-trifluoro-5-nitro, pubchem1071, acmc-1baej, 3,4,5-trifluronitrobenzene, ksc495s3n, 3,4,5-trifluoronitro benzene
PubChem CID 2782793
IUPAC Name 1,2,3-trifluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(F)C(F)=C1

Specifications

Density 1.52
Flash Point 75°C (167°F)
Refractive Index 1.486
Quantity 5 g
UN Number UN2810
Beilstein 2455018
Formula Weight 177.08
Percent Purity 99%
Chemical Name or Material 1,2,3-Trifluoro-5-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.