Learn More
1,2,4,5-Tetrachlorobenzene, TRC
Product Code. 30179928
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179928 50 g 50g
30180246 10 g 10g
30179735 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179928 Supplier Toronto Research Chemicals Supplier No. TRCT29128550G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95-94-3
Molecular Formula C6 H2 Cl4
Molecular Weight (g/mol) 215.89
Synonym Benzene, 1,2,4,5-tetrachloro-, 1,2,4,5-Tetrachlorobenzene, NSC 27003, s-Tetrachlorobenzene
IUPAC Name 1,2,4,5-tetrachlorobenzene
SMILES Clc1cc(Cl)c(Cl)cc1Cl
Specifications
Quantity 50 g
InChI Formula InChI=1S/C6H2Cl4/c7-3-1-4(8)6(10)2-5(3)9/h1-2H
Formula Weight 213.8911
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene