Learn More
1-(2,4,6-Trihydroxyphenyl)-2-(4-methoxyphenyl)ethanone, TRC
Product Code. 30454632
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454632 100 mg 100mg
30452584 50 mg 50mg
30454315 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30454632 Supplier Toronto Research Chemicals Supplier No. TRCT895800100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15485-66-2
Molecular Formula C15H14O5
Molecular Weight (g/mol) 274.26
IUPAC Name 2-(4-methoxyphenyl)-1-(2,4,6-trihydroxyphenyl)ethanone
SMILES COc1ccc(CC(=O)c2c(O)cc(O)cc2O)cc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H14O5/c1-20-11-4-2-9(3-5-11)6-12(17)15-13(18)7-10(16)8-14(15)19/h2-5,7-8,16,18-19H,6H2,1H3
Formula Weight 274.0841
Chemical Name or Material 1-(2,4,6-Trihydroxyphenyl)-2-(4-methoxyphenyl)ethanone