missing translation for 'onlineSavingsMsg'
Learn More

1,2,4-Trichloro-5-iodobenzene, 98%, Thermo Scientific™

Product Code. 11392326
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392326 5 g 5g
11302336 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392326 Supplier Thermo Scientific Alfa Aesar Supplier No. L06015.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,2,4-Trichloro-5-iodobenzene can be used to produce 3-(2,4,5-trichloro-phenyl)-acrylic acid tert-butyl ester at the temperature of 80°C.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in the dark. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7145-82-6
Molecular Formula C6H2Cl3I
Molecular Weight (g/mol) 307.336
MDL Number MFCD00051450
InChI Key KKXMBRLCXIPURK-UHFFFAOYSA-N
Synonym 1-iodo-2,4,5-trichlorobenzene, 2,4,5-trichloroiodobenzene, benzene,2,4-trichloro-5-iodo, attercop-chm at126309, benzene,1,2,4-trichloro-5-iodo, 1,2,4-tris chloranyl-5-iodanyl-benzene
PubChem CID 252788
IUPAC Name 1,2,4-trichloro-5-iodobenzene
SMILES C1=C(C(=CC(=C1Cl)I)Cl)Cl

Specifications

Melting Point 102°C to 105°C
Boiling Point 294°C (110 mmHg)
Quantity 5 g
Beilstein 1940881
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 307.35
Percent Purity 98%
Chemical Name or Material 1,2,4-Trichloro-5-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.