missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Dimethylphenyl)-1,5,6,7-tetrahydro-2H-azepin-2-one, TRC

Product Code. 30133602
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133602 100 mg 100mg
30006974 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133602 Supplier Toronto Research Chemicals Supplier No. TRCD477200100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797894-80-4
Molecular Formula C14H17NO
Molecular Weight (g/mol) 215.29
Synonym Levobupivacaine Impurity C;
SMILES CC1=C(C(=CC=C1)C)N2CCCC=CC2=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H17NO/c1-11-7-6-8-12(2)14(11)15-10-5-3-4-9-13(15)16/h4,6-9H,3,5,10H2,1-2H3
Formula Weight 215.131
Percent Purity >95%
Chemical Name or Material 1-(2,6-Dimethylphenyl)-1,5,6,7-tetrahydro-2H-azepin-2-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.