missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzene-d4-diamine, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,2-Phenylenediamine-d4 |
| CAS | 291765-93-0 |
| Molecular Formula | C6 D4 H4 N2 |
| Quantity | 1 g |
| Synonym | 1,2-Benzenediamine-d4, 1,2-Benzene-d4-diamine, o-Phenylenediamine-d4, 1,2-Diaminobenzene-d4, 1,2-Benzene-3,4,5,6-d4-diamine, 3,4,5,6-Tetradeuteriobenzene-1,2-diamine, 2-Aminoaniline-d4, C.I. 76010-d4, C.I. Oxidation Base 16-d4, IK 3, NSC 5354-d4, Orthamine-d4, o-Aminoaniline-d4, o-Aminophenylamine-d4, o-Benzenediamine-d4, o-Diaminobenzene-d4, o-Phenylenediamine-d4, 1,2-Diaminobenzene-3,4,5,6-d4, Benzene-1,2-diamine-D4 |
| InChI Formula | InChI=1 S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4 H,7-8H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N)c(N)c1[2 H] |
| IUPAC Name | 3,4,5,6-tetradeuteriobenzene-1,2-diamine |
| Molecular Weight (g/mol) | 112.17 |
| Formula Weight | 112.0939 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction