Learn More
1,2-Bis(4-methoxyphenyl)ethanone, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Bis(4-methoxyphenyl)ethanone |
| CAS | 120-44-5 |
| Molecular Formula | C16H16O3 |
| Quantity | 5 g |
| Synonym | 4'-Methoxy-2-(p-methoxyphenyl)-acetophenone, 1,2-Bis(4-methoxyphenyl)ethanone, 1,2-Bis(p-methoxyphenyl)ethanone, 1,2-Di-p-anisylethanone, 4,4'-Dimethoxydeoxybenzoin, 4-Methoxybenzyl 4-methoxyphenyl ketone, 4'-Methoxy-2-(4-methoxyphenyl)acetophenone, 4'-Methoxy-2-(p-methoxyphenyl)acetophenone, Deoxy-p-anisoin, Deoxyanisoin, Desoxy-p-anisoin, NSC 26660, NSC 8793, p-Methoxybenzyl p-methoxyphenyl ketone, α-(p-Methoxyphenyl)-p-methoxyacetophenone |
| InChI Formula | InChI=1S/C16H16O3/c1-18-14-7-3-12(4-8-14)11-16(17)13-5-9-15(19-2)10-6-13/h3-10H,11H2,1-2H3 |
| SMILES | O=C(C1=CC=C(OC)C=C1)CC2=CC=C(OC)C=C2 |
| IUPAC Name | 1,2-bis(4-methoxyphenyl)ethan-1-one |
| Molecular Weight (g/mol) | 256.3 |
| Formula Weight | 256.11 |