Learn More
1,2-Di(furan-2-yl)ethane-1,2-dione, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 492-94-4 |
| Molecular Formula | C10H6O4 |
| MDL Number | MFCD00003241 |
| Quantity | 5 g |
| SMILES | O=C(C1=CC=CO1)C(C2=CC=CO2)=O |
| Molecular Weight (g/mol) | 190.15 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,2-Di(furan-2-yl)ethane-1,2-dione |