missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diamino-4,5-methylenedioxybenzene, Dihydrochloride, TRC

Product Code. 30010199
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010199 10 mg 10mg
30012184 25 mg 25mg
30004484 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010199 Supplier Toronto Research Chemicals Supplier No. TRCD41637010MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81864-15-5
Molecular Formula C7 H8 N2 O2 . 2 Cl H
Molecular Weight (g/mol) 225.07
Synonym 1,3-Benzodioxole-5,6-diamine, hydrochloride (1:2), 1,3-Benzodioxole-5,6-diamine, dihydrochloride (9CI), 1,3-Benzodioxole-5,6-diamine hydrochloride (1:2), 1,2-Diamino-4,5-methylenedioxybenzene dihydrochloride, 4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride
IUPAC Name 1,3-benzodioxole-5,6-diamine;dihydrochloride
SMILES Cl.Cl.Nc1cc2OCOc2cc1N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H8N2O2.2ClH/c8-4-1-6-7(2-5(4)9)11-3-10-6;;/h1-2H,3,8-9H2;2*1H
Formula Weight 224.0119
Percent Purity >95%
Chemical Name or Material 1,2-Diamino-4,5-methylenedioxybenzene, Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.