Learn More
1,2-Diaminopropane-N,N,N',N'-tetraacetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Diaminopropane-N,N,N',N'-tetraacetic acid |
| CAS | 4408-81-5 |
| Molecular Formula | C11 H18 N2 O8 |
| Quantity | 10 g |
| Synonym | Acetic acid, (propylenedinitrilo)tetra- (6CI,7CI,8CI), N,N'-(1-Methyl-1,2-ethanediyl)bis[N-(carboxymethyl)glycine], (±)-1,2-Diaminopropanetetraacetic acid, 1,2-Diaminopropane-N,N,N',N'-tetraacetic acid, 1,2-Diaminopropanetetraacetate, 1,2-Diaminopropanetetraacetic acid, 1,2-Propanediaminetetraacetic acid, 1,2-Propylenediamine-N,N,N',N'-tetraacetic acid, 1,2-Propylenediaminetetraacetic acid, 2,2',2'',2'''-(1,2-Propanediyldinitrilo)tetraacetic acid, DL-Propylenediaminetetraacetic acid, Dissolvine P, Dissolvine PDZ, MeEDTA, Methyl-EDTA, Propylene-1,2-diaminetetraacetic acid, Trilon FS |
| InChI Formula | InChI=1S/C11H18N2O8/c1-7(13(5-10(18)19)6-11(20)21)2-12(3-8(14)15)4-9(16)17/h7H,2-6H2,1H3,(H,14,15)(H,16,17)(H,18,19)(H,20,21) |
| SMILES | CC(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O |
| IUPAC Name | 2-[2-[bis(carboxymethyl)amino]propyl-(carboxymethyl)amino]acetic acid |
| Molecular Weight (g/mol) | 306.27 |
| Formula Weight | 306.1063 |