missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibenzoylethylene, predominantly trans, 96%

Product Code. 11353286
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353286 5 g 5g
11343286 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353286 Supplier Thermo Scientific Alfa Aesar Supplier No. L12643.06
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 959-28-4 | C16H12O2 | 236.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959-28-4
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00003083
InChI Key WYCXGQSQHAXLPK-VAWYXSNFSA-N
Synonym trans-1,2-dibenzoylethylene, dibenzoylethylene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, trans-1,4-diphenyl-2-butene-1,4-dione, 1,2-dibenzoylethene, ethylene, 1,2-dibenzoyl, usaf nd-57, 2-butene-1,4-dione, 1,4-diphenyl, trans-diphenacylidene
PubChem CID 735960
IUPAC Name (E)-1,4-diphenylbut-2-ene-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2

Specifications

Melting Point 103°C to 108°C
Quantity 5 g
Beilstein 743958
Formula Weight 236.27
Percent Purity 96%
Chemical Name or Material 1,2-Dibenzoylethylene, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.