missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrafluorobenzene, 99%

Product Code. 11417967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417967 1 g 1g
11427967 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417967 Supplier Thermo Scientific Alfa Aesar Supplier No. A16620.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 827-08-7 | C6Br2F4 | 307.868 g/mol

1,2-Dibromotetrafluorobenzene is used as medicine and liquid crystal material intermediate. It helps in the synthesis of diphenyl(o-bromotetrafluorophenyl)phosphine and 2,3,4,5tetrafluorothioanisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromotetrafluorobenzene is used as medicine and liquid crystal material intermediate. It helps in the synthesis of diphenyl(o-bromotetrafluorophenyl)phosphine and 2,3,4,5tetrafluorothioanisole.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-08-7
Molecular Formula C6Br2F4
Molecular Weight (g/mol) 307.868
MDL Number MFCD00039209
InChI Key IPLUWQPTPKNBRD-UHFFFAOYSA-N
Synonym 1,2-dibromotetrafluorobenzene, 3,4,5,6-tetrafluoro-1,2-dibromobenzene, tetrafluoro-1,2-dibromobenzene, benzene, 1,2-dibromotetrafluoro, benzene, 1,2-dibromo-3,4,5,6-tetrafluoro, 1,2-dibromoperfluorobenzene, dtfb, pubchem2195, dibromotetrafluorobenzene, acmc-20ap6i
PubChem CID 70007
IUPAC Name 1,2-dibromo-3,4,5,6-tetrafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)Br)Br)F)F)F

Specifications

Density 2.26
Boiling Point 197°C to 199°C
Flash Point 99°C (210°F)
Refractive Index 1.516
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 307.87
Percent Purity 99%
Chemical Name or Material 1,2-Dibromotetrafluorobenzene
compliance-icons
Product Identifier
  • 1,2-Dibromotetrafluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,2-Dibromo-3,4,5,6-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.