missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Dichloropropane-d6, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dichloropropane-d6 |
| CAS | 93952-08-0 |
| Molecular Formula | C3 D6 Cl2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C3H6Cl2/c1-3(5)2-4/h3H,2H2,1H3/i1D3,2D2,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Cl)C([2 H])([2 H])Cl |
| IUPAC Name | 1,2-dichloro-1,1,2,3,3,3-hexadeuterio-propane |
| Molecular Weight (g/mol) | 119.02 |
| Formula Weight | 118.0223 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction