missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dimethoxybenzene-d10, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,2-Dimethoxybenzene D10 |
| CAS | 362049-43-2 |
| Molecular Formula | C8 D10 O2 |
| Quantity | 0.5 g |
| Synonym | Benzene-1,2,3,4-d4, 5,6-di(methoxy-d3)-, 5,6-Di(methoxy-d3)benzene-1,2,3,4-d4, 1,2-Dimethoxybenzene D10, 1,2-Dimethoxybenzene-d10, 2-Methoxyanisole-d10, Catechol-d4 di(methyl-d3) ether, o-Hydroquinone-d4 di(methyl-d3) ether |
| InChI Formula | InChI=1 S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(OC([2 H])([2 H])[2 H])c(OC([2 H])([2 H])[2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4-tetradeuterio-5,6-bis(trideuteriomethoxy)benzene |
| Molecular Weight (g/mol) | 148.225 |
| Formula Weight | 148.131 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction