missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dipiperidinoethane, 98%

Product Code. 10792611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10792611 1 g 1g
10646392 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792611 Supplier Thermo Scientific Acros Supplier No. 185110010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1932-04-3 | C12H24N2 | 196.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-3-1932
Molecular Formula C12H24N2
Molecular Weight (g/mol) 196.33
MDL Number MFCD00014644
InChI Key UYMQPNRUQXPLCY-UHFFFAOYSA-N
Synonym 1,2-dipiperidinoethane, 1,2-dipiperidinylethane, piperidine, 1,1'-1,2-ethanediyl bis, 1-2-piperidin-1-ylethyl piperidine, 1,2-di piperidin-1-yl ethane, 1,2-bis piperidino ethane, piperidine, 1,1'-ethylenedi, 1-2-piperidin-1-yl ethyl piperidine, 1,1'-ethylenedipiperidine, piperidine,1'-ethylenedi
PubChem CID 74733

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1°C to 0.5°C
Density 0.9100g/mL
Boiling Point 265°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4865 to 1.4885
Quantity 1 g
Beilstein 20, I, 19
Specific Gravity 0.91
Formula Weight 196.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 2-Dipiperidinoethane, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.