missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dipiperidinoethane (DPE), Mikromol™

Product Code. 30052742
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052742 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052742 Supplier Mikromol Supplier No. MM0486.02
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,2-Dipiperidinoethane (DPE)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dipiperidinoethane
CAS 4-3-1932
Analyte or Component Names 1,2-Dipiperidinoethane
Shipping Condition Room Temperature
Molecular Formula C12 H24 N2
Certifications/Compliance ISO 9001
Synonym 1,2-Dipiperidinoethane, DPE
InChI Formula InChI=1S/C12H24N2/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12H2
SMILES C(CN1CCCCC1)N2CCCCC2
IUPAC Name 1-(2-piperidin-1-ylethyl)piperidine
Molecular Weight (g/mol) 196.33
Quantity 100 mg
Formula Weight 196.1939 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.