missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-3,3,3-trifluoropropane, 98%

Product Code. 11322317
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322317 1 g 1g
11332317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322317 Supplier Thermo Scientific Alfa Aesar Supplier No. L09869.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 359-41-1 | C3H3F3O | 112.051 g/mol

1,2-Epoxy-3,3,3-trifluoropropane is used for the synthesis of trifluorolactic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-3,3,3-trifluoropropane is used for the synthesis of trifluorolactic acid. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Not miscible with water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible materials are heat, oxidizing agents and water. Store away from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 359-41-1
Molecular Formula C3H3F3O
Molecular Weight (g/mol) 112.051
MDL Number MFCD00041506
InChI Key AQZRARFZZMGLHL-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-2,3-epoxypropane, 2-trifluoromethyl oxirane, 3,3,3-trifluoro-1,2-epoxypropane, 1,2-epoxy-3,3,3-trifluoropropane, oxirane, trifluoromethyl, 3,3,3-trifluoropropane epoxide, pubchem20840, trifluoromethyl oxirane, acmc-20apk3
PubChem CID 520769
IUPAC Name 2-(trifluoromethyl)oxirane
SMILES C1C(O1)C(F)(F)F

Specifications

Density 1.3
Boiling Point 38°C to 40°C
Flash Point −40°C (−40°F)
Refractive Index 1.297
Quantity 1 g
UN Number UN1992
Beilstein 103730
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 112.05
Percent Purity 98%
Chemical Name or Material 1,2-Epoxy-3,3,3-trifluoropropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.