Learn More
1-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethan-1-one, 95%, BLD Pharm™
Product Code. 31088262
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31088262 10 g
31082355 250 mg
31076651 1 g
31084913 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31088262 Supplier BLD Pharm Supplier No. BD0081174010g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 1351499-39-2
Molecular Formula C14H18BFO3
MDL Number MFCD18732911
Quantity 10 g
SMILES CC(C1=CC=C(B2OC(C)(C)C(C)(C)O2)C=C1F)=O
Molecular Weight (g/mol) 264.10
Percent Purity 95%
Chemical Name or Material 1-(2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethan-1-one