Learn More
1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline, TRC
Product Code. 30339854
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339854 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30339854 Supplier Toronto Research Chemicals Supplier No. TRCB43640550MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(2-Fluorophenyl)-1,2,3,4-tetrahydroisoquinoline
CAS 1017125-85-7
Molecular Formula C15H14NF
Quantity 50 mg
InChI Formula InChI=1S/C15H14FN/c16-14-8-4-3-7-13(14)15-12-6-2-1-5-11(12)9-10-17-15/h1-8,15,17H,9-10H2
SMILES Fc1ccccc1C2NCCc3ccccc23
IUPAC Name 1-(2-fluorophenyl)-1,2,3,4-tetrahydroisoquinoline
Molecular Weight (g/mol) 227.28