missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)piperazine, 97%

Product Code. 10501121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10501121 10 g Plastic bottle 10g
10511121 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10501121 Supplier Thermo Scientific Acros Supplier No. 268690100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 1011-15-0 | C10H13FN2 | 180.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1011-15-0
Molecular Formula C10H13FN2
Molecular Weight (g/mol) 180.22
MDL Number MFCD00040726
InChI Key IVTZRJKKXSKXKO-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl piperazine, 1-2-fluorophenyl-piperazine, 1-2-fluoro-phenyl-piperazine, piperazine, 1-2-fluorophenyl, chembl9429, 2-fluorophenyl piperazine, ofpp, pubchem8573, 1-2-fluorophenyl piperazinehydrochloride, 2-fluorophenylpiperazine
PubChem CID 70529
IUPAC Name 1-(2-fluorophenyl)piperazine
SMILES C1CN(CCN1)C2=CC=CC=C2F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1410g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Refractive Index 1.5550 to 1.558
Quantity 10 g
Specific Gravity 1.141
Formula Weight 180.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(2-Fluorophenyl)piperazine, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(2-Fluorophenyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.