missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1,2-Hexanediol-1,1,2-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | (+/-)-1,2-Hexanediol-1,1,2-D3 |
| Molecular Formula | C6 D3 H11 O2 |
| Quantity | 0.1 g |
| Synonym | Hexane-1,2-diol-1,1,2-d3, 1,2-Dihydroxyhexane-1,1,2-d3, (±)-1,2-Hexanediol-1,1,2-d3, (±)-1,2-Hexanediol-1,1,2-D3, 1,2-Hexanediol-d3 |
| InChI Formula | InChI=1 S/C6H14O2/c1-2-3-4-6(8)5-7/h6-8 H,2-5H2,1H3/i5D2,6 D |
| SMILES | [2 H]C([2 H])(O)C([2 H])(O)CCCC |
| IUPAC Name | 1,1,2-trideuteriohexane-1,2-diol |
| Molecular Weight (g/mol) | 121.1927 |
| Formula Weight | 121.1182 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction