missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Hydroxy-5-methylphenyl)ethanone, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1-(2-Hydroxy-5-methylphenyl)ethanone |
| CAS | 1450-72-2 |
| Molecular Formula | C9H10O2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C9H10O2/c1-6-3-4-9(11)8(5-6)7(2)10/h3-5,11H,1-2H3 |
| SMILES | CC(=O)c1cc(C)ccc1O |
| IUPAC Name | 1-(2-hydroxy-5-methylphenyl)ethanone |
| Molecular Weight (g/mol) | 150.17 |
| Formula Weight | 150.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction