Learn More
1-(2-Methoxyphenyl)piperazine Hydrobromide, TRC
Product Code. 30479070
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479070 500 mg 500mg
30477165 100 mg 100mg
30465725 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30479070 Supplier Toronto Research Chemicals Supplier No. TRCM226563500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100939-96-6
Molecular Formula C11H16N2O . HBr
Molecular Weight (g/mol) 273.17
IUPAC Name 1-(2-methoxyphenyl)piperazine;hydrobromide
SMILES Br.COc1ccccc1N2CCNCC2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C11H16N2O.BrH/c1-14-11-5-3-2-4-10(11)13-8-6-12-7-9-13;/h2-5,12H,6-9H2,1H3;1H
Formula Weight 272.0524
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine Hydrobromide