missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Methoxyphenyl)piperazine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-(2-Methoxyphenyl)Piperazine |
| CAS | 35386-24-4 |
| Molecular Formula | C11 H16 N2 O |
| Quantity | 5 g |
| Synonym | 1-(2-Methoxyphenyl)piperazine, 1-(o-Anisyl)piperazine, 1-(o-Methoxyphenyl)piperazine, 1-[2-(Methyloxy)phenyl]piperazine, D 15157, N-(2-Methoxyphenyl)piperazine, N-(o-Methoxyphenyl)piperazine, o-Methoxyphenylpiperazine |
| InChI Formula | InChI=1S/C11H16N2O/c1-14-11-5-3-2-4-10(11)13-8-6-12-7-9-13/h2-5,12H,6-9H2,1H3 |
| SMILES | COc1ccccc1N2CCNCC2 |
| IUPAC Name | 1-(2-methoxyphenyl)piperazine |
| Molecular Weight (g/mol) | 192.26 |
| Formula Weight | 192.1263 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction